missing translation for 'onlineSavingsMsg'
Learn More
Mercuric acetate, 98%, Thermo Scientific Chemicals Product Code.: 10327740

Mercuric acetate, 98%, Thermo Scientific Chemicals

Product Code. 10327740
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Product Code. 10327740

Brand: Thermo Scientific Acros 175020050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
MDL Number MFCD00012165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179.0°C to 182.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 98%
Molecular Formula C4H6HgO4
Linear Formula (CH3CO2)2Hg
Beilstein 02, 96
Fieser 01,644; 02,264; 03,194; 04,319; 05,424; 06,358; 10,252; 11,315; 12,298; 17,176
Merck Index 15, 5940
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 318.68
Formula Weight 318.67
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Mercuric acetate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0434749 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number175021000,175020025,175025000,17502001,17502048,175020050
Lot NumberA0456175 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number175021000,175020025,175025000,17502001,17502048,175020050
Lot NumberA0454364 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number175021000,175020025,175025000,17502001,17502048,175020050
Lot NumberA0445692 Certificate TypeCertificate of Analysis Date07/09/2022 Catalog Number175021000,175020025,175025000,17502001,17502048,175020050
Lot NumberA0394180 Certificate TypeCertificate of Analysis Date23/10/2020 Catalog Number175021000,175020025,175025000,17502001,17502048,175020050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercuric acetate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.