missing translation for 'onlineSavingsMsg'
Learn More
Mercury(Ii) Chloride, ACS Reagent, Honeywell Fluka™ Product Code.: 15691990

Mercury(Ii) Chloride, ACS Reagent, Honeywell Fluka™

Product Code. 15691990 Shop All Honeywell Fluka Products
500 g
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15691990

Brand: Honeywell-Fluka 215465500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2Show MoreShow Less
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Quantity 500 g
Linear Formula HgCl2
UN Number 1624
Solubility Information Soluble in diethyl ether, acetic acid, ethyl acetate, glycerol, alcohol, acetone (salt form), water
Formula Weight 271.5g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Vapor Pressure 1.3mmHg (236°C)
Chemical Name or Material Mercury(Ii) Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(Ii) Chloride, ACS Reagent, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.