missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) Chloride, Certified AR for Analysis, meets analytical specification of BP, Fisher Chemical™
Click to view available options
Quantity:
250 g
Specifications
Specifications
Boiling Point | 302°C |
Melting Point | 277°C |
Color | White |
pH | 3.3 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 271.5g/mol |
Packaging | Plastic powder jar |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercury(II) Chloride, Certified AR for Analysis, meets analytical specification of BP, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction