missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) selenide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11368107

Mercury(II) selenide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11368107
2 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11368107

Brand: Thermo Scientific Alfa Aesar 022878.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20601-83-6
Molecular Formula HgSe
Molecular Weight (g/mol) 279.56
MDL Number MFCD00049743
InChI Key YQMLDSWXEQOSPP-UHFFFAOYSA-N
Synonym mercury selenide, mercury ii selenide, mercury-selenium complex, mercury monoselenide, mercury 2+ selenide, mercury ii selenide trace metals basis 1gShow MoreShow Less
PubChem CID 88609
IUPAC Name selanylidenemercury
SMILES [Se]=[Hg]

Specifications

Melting Point (sublimation)
Color Gray
Physical Form Powder
Quantity 2 g
UN Number UN2025
Formula Weight 279.55
Percent Purity 99.9%
Odor Odorless
Density 8.21 g/mL
Assay (metals basis)
Chemical Name or Material Mercury(II) selenide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC13L13 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number022878.09,022878.04
Lot NumberW18A046 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number022878.09,022878.04
Lot NumberI11K14 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number022878.09,022878.04
Lot NumberG22W003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number022878.09,022878.04
Lot NumberC02X012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number022878.09,022878.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) selenide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.