missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) thiocyanate, 99+%, Thermo Scientific Chemicals Product Code.: 10296440

Mercury(II) thiocyanate, 99+%, Thermo Scientific Chemicals

Product Code. 10296440
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10296440

Brand: Thermo Scientific Acros 196890500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-85-8
Molecular Formula C4HgK2N4S4
Molecular Weight (g/mol) 511.10
MDL Number MFCD00011039
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N

Specifications

Melting Point 165.0°C
Color White
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula Hg(SCN)2
Fieser 09,293
Flash Point 120°C
Merck Index 15, 5958
Solubility Information Solubility in water: 0.7g/L (25°C). Other solubilities: soluble in diluted hcl
Formula Weight 316.75
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Mercury(II) thiocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467030 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number196890500,196892500
Lot NumberA0459132 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number196890500,196892500
Lot NumberA0458102 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number196890500,196892500
Lot NumberA0449335 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog Number
Lot NumberA0400542 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number196890500,196892500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) thiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P330-Rinse mouth.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) thiocyanate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.