missing translation for 'onlineSavingsMsg'
Learn More
meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals Product Code.: 11496816

meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals

Product Code. 11496816
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11496816

Brand: Thermo Scientific Alfa Aesar B23946.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

Solubility
Soluble in hot alcohol or chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064253
InChI Key IHPDTPWNFBQHEB-UHFFFAOYNA-N
Synonym meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb componentShow MoreShow Less
PubChem CID 853018
ChEBI CHEBI:50015
IUPAC Name (1R,2S)-1,2-diphenylethane-1,2-diol
SMILES OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 134°C to 136°C
Quantity 25 g
Beilstein 2050814
Merck Index 14,4777
Solubility Information Soluble in hot alcohol or chloroform.
Formula Weight 214.27
Percent Purity 99%
Chemical Name or Material meso-Hydrobenzoin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW12K013 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog NumberB23946.06,B23946.14
Lot Number10149380 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23946.06,B23946.14
Lot NumberQ11F024 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23946.06,B23946.14
Lot NumberY26F029 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23946.06,B23946.14
Lot NumberY27F044 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23946.06,B23946.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.