missing translation for 'onlineSavingsMsg'
Learn More
Metaldehyde, 99%, Thermo Scientific Chemicals Product Code.: 10371034

Metaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 10371034
1 kg, Plastic bottle
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10371034

Brand: Thermo Scientific Acros 271860010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-62-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00071549
InChI Key GKKDCARASOJPNG-UHFFFAOYSA-N
Synonym metaldehyde, metacetaldehyde, cekumeta, ariotox, halizan, metason, acetaldehyde, tetramer, slug-tox, corry's slug death, metaldehydShow MoreShow Less
PubChem CID 61021
ChEBI CHEBI:81931
IUPAC Name 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
SMILES CC1OC(OC(OC(O1)C)C)C

Specifications

Melting Point 246.0°C
Color White
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3CHO)4
Quantity 1 kg
Beilstein 01, 602
Merck Index 14, 5923
Solubility Information Solubility in water: 0.02 w/w % (20°C). Other solubilities: soluble in benzene,chloroform,sparingly soluble in alcohol,ether
Sublimation Point 0112.0°C
Formula Weight 176.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Metaldehyde, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458770 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog Number271862500,271860010,271860050
Lot NumberA0453631 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number271862500,271860010,271860050
Lot NumberA0452940 Certificate TypeCertificate of Analysis Date10/05/2023 Catalog Number271862500,271860010,271860050
Lot NumberA0403056 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number271862500,271860010,271860050
Lot NumberA0407184 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number271862500,271860010,271860050
compliance-icons compliance-icons
Product Identifier
  • Metaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable solid Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metaldehyde, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.