missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, ACS reagent, ≥99.8%, Honeywell Riedel-de Haën™
ACS reagent,≥99.8%
£34.31 - £338.89
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow More |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15671610
|
Honeywell-Riedel-de Haen
179337-500ML |
500 mL |
£34.31
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15641610
|
Honeywell-Riedel-de Haen
179337-1L |
1 L | N/A | ||||||
15651610
|
Honeywell-Riedel-de Haen
179337-2.5L |
2.5 L | N/A | ||||||
15691610
|
Honeywell-Riedel-de Haen
179337-6X500ML |
6 x 500 mL |
£168.27
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15678320
|
Honeywell-Riedel-de Haen
179337-4L |
4 L | N/A | ||||||
15681610
|
Honeywell-Riedel-de Haen
179337-6X1L |
6 x 1 L | N/A | ||||||
15661610
|
Honeywell-Riedel-de Haen
179337-20L |
20 L | N/A | ||||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow More | |
CHEBI:17790 | |
CO |
Specifications
-98°C (literature) | |
100.0000% | |
64.7°C | |
Glass Bottle | |
CH3OH | |
UN1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
>99.8% | |
Methanol, ACS reagent, ≥99.8% |
67-56-1 | |
0.791 g/mL at 25 °C(lit.) | |
11°C | |
CH4O | |
MFCD00004595 | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
ACS Reagent |
Safety and Handling
missing translation for 'shelfLife' : 547 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
missing translation for 'einecsNumber' : 200-659-6
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanol, ACS reagent, ≥99.8%, Honeywell Riedel-de Haën™