missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For Moisture Titration
Specifications
Specifications
Quantity | 1 L |
Flash Point | 12.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.80kg |
Formula Weight | 32.04 |
Grade | Meets ACS reagent chemical requirements |
Chemical Name or Material | Methanol, Anhydrous |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Safety and Handling



Signal Word
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction