missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Methionine/Choline Deficient Diet (Powdered)

Methionine/Choline Deficient Diet (Powdered) has been used to study nonalcoholic steatohepatitis (NASH).

Brand:  MP Biomedicals™ 0296043810

 View more versions of this product

Product Code. 11405365

  • £1355.00 / 10kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains sucrose, corn starch, corn oil, Alphacel non-nutritive bulk, and AIN 76 mineral mix.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Powder
10 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.