missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Methionine/Choline Deficient Diet (Powdered)
Methionine/Choline Deficient Diet (Powdered) has been used to study nonalcoholic steatohepatitis (NASH).
£1355.00
Description
- Contains sucrose, corn starch, corn oil, Alphacel non-nutritive bulk, and AIN 76 mineral mix.
Specifications
Powder |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title