missing translation for 'onlineSavingsMsg'
Learn More
Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%, Thermo Scientific™ Product Code.: 15468685

Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15468685
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15468685

Brand: Thermo Scientific Alfa Aesar H33887.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184174-81-0
Molecular Formula C11H15NO2S
Molecular Weight (g/mol) 225.306
MDL Number MFCD01114967
InChI Key BDPLKEMLVDUYRR-UHFFFAOYSA-N
Synonym methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylate, methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta-b thiophene-3-carboxylate, methyl 2-amino-4h,5h,6h,7h,8h-cyclohepta b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, 5,6,7,8-tetrahydro-2-amino-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, methyl 2-amino-4,5,6,7,8-pentahydrocyclohepta 1,2-b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b-thiophene-3-carboxylic acid methyl esterShow MoreShow Less
PubChem CID 2756554
IUPAC Name methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
SMILES COC(=O)C1=C(SC2=C1CCCCC2)N

Specifications

Melting Point 95°C to 96°C
Quantity 5 g
Formula Weight 225.31
Percent Purity 96%
Chemical Name or Material Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10172142 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.