missing translation for 'onlineSavingsMsg'
Learn More
Methyl 2-chloropropionate, 97%, Thermo Scientific Chemicals Product Code.: 11418467

Methyl 2-chloropropionate, 97%, Thermo Scientific Chemicals

Product Code. 11418467
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11418467

Brand: Thermo Scientific Alfa Aesar A18541.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

Solubility
Insoluble in water

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17639-93-9
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
MDL Number MFCD00000863
InChI Key JLEJCNOTNLZCHQ-UHFFFAOYNA-N
Synonym methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6fShow MoreShow Less
PubChem CID 28659
IUPAC Name methyl 2-chloropropanoate
SMILES COC(=O)C(C)Cl

Specifications

Melting Point <-10°C
Density 1.14
Boiling Point 132°C to 133°C
Flash Point 38°C (100°F)
Refractive Index 1.417
Quantity 250 g
UN Number UN2933
Beilstein 1421535
Solubility Information Insoluble in water
Formula Weight 122.55
Percent Purity 97%
Chemical Name or Material Methyl 2-chloropropionate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM12H001 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18541.30,A18541.0B
Lot NumberT21G036 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18541.30,A18541.0B
Lot Number10000624 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18541.30,A18541.0B
Lot Number14824 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18541.30,A18541.0B
Lot Number10005608 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18541.30,A18541.0B
compliance-icons compliance-icons
Product Identifier
  • Methyl 2-chloropropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 2-chloropropionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 2-chloropropionate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.