missing translation for 'onlineSavingsMsg'
Learn More
Methyl 3,5-dibromo-2-hydroxybenzoate, 98%, Thermo Scientific Chemicals Product Code.: 11466067

Methyl 3,5-dibromo-2-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11466067
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11466067

Brand: Thermo Scientific Alfa Aesar B25077.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21702-79-4
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00017534
InChI Key KISQISIKTRRMOX-UHFFFAOYSA-N
PubChem CID 624217
IUPAC Name methyl 3,5-dibromo-2-hydroxybenzoate
SMILES COC(=O)C1=CC(Br)=CC(Br)=C1O

Specifications

Melting Point 150°C to 152°C
Quantity 25 g
Synonym methyl 3,5-dibromosalicylate, benzoic acid, 3,5-dibromo-2-hydroxy-, methyl ester, 3,5-dibromo-2-hydroxybenzoic acid methyl ester, pubchem3976, acmc-1cpm5, rarechem al bf 0030, timtec-bb sbb001273, 3,5-dibromo-2-hydroxymethyl benzoate, benzoic acid,3,5-dibromo-2-hydroxy-, methyl ester
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromo-2-hydroxybenzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ8282A Certificate TypeCertificate of Analysis Date05/05/2023 Catalog NumberB25077.22,B25077.14
Lot NumberJ8282A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25077.22,B25077.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 3,5-dibromo-2-hydroxybenzoate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.