missing translation for 'onlineSavingsMsg'
Learn More
Methyl carbazate, 98%, Thermo Scientific Chemicals Product Code.: 11461928

Methyl carbazate, 98%, Thermo Scientific Chemicals

Product Code. 11461928
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11461928

Brand: Thermo Scientific Alfa Aesar A14548.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl carbazate was used in the preparation of methyl 3-(4-methyl-benzyl-idene)carbazate. It was also used in the synthesis of imidazo[1,5-d][1,2,4]triazines. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl carbazate was used in the preparation of methyl 3-(4-methyl-benzyl-idene)carbazate. It was also used in the synthesis of imidazo[1,5-d][1,2,4]triazines. It is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6294-89-9
Molecular Formula C2H6N2O2
Molecular Weight (g/mol) 90.08
MDL Number MFCD00007594
InChI Key WFJRIDQGVSJLLH-UHFFFAOYSA-N
Synonym methyl carbazate, methyl hydrazinecarboxylate, methyl hydrazinocarboxylate, methoxycarbohydrazide, methylcarbazate, hydrazinecarboxylic acid, methyl ester, carbomethoxyhydrazide, methyl hydrazinoformate, methoxycarbonyl hydrazine, carbazic acid methyl esterShow MoreShow Less
PubChem CID 80519
IUPAC Name methyl N-aminocarbamate
SMILES COC(=O)NN

Specifications

Melting Point 70°C to 73°C
Odor Weak
Quantity 2500 g
UN Number UN2811
Beilstein 1679395
Solubility Information Soluble in water.
Formula Weight 90.08
Percent Purity 98%
Chemical Name or Material Methyl carbazate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244901 Certificate TypeCertificate of Analysis Date05/12/2022 Catalog NumberA14548.0E,A14548.22
Lot Number0900A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14548.0E,A14548.22
Lot Number10235876 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14548.0E,A14548.22
Lot NumberGBFA021454 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14548.0E,A14548.22
Lot Number10097603 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14548.0E,A14548.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl carbazate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.