missing translation for 'onlineSavingsMsg'
Learn More
Methyl formate, 98%, for spectroscopy, Thermo Scientific Chemicals Product Code.: 10445851

Methyl formate, 98%, for spectroscopy, Thermo Scientific Chemicals

Product Code. 10445851
500 mL, Glass bottle
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10445851

Brand: Thermo Scientific Acros 414345000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3Show MoreShow Less
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Acidity 0.0008 meq/g max.
Absorbance 0.009 max. at 320nm, 0.032 max. at 300nm, 0.051 max. at 270nm, 1 max. at 260nm
Melting Point -100.0°C
Density 0.9680g/mL
Boiling Point 31.0°C to 33.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HCO2CH3
Refractive Index 1.3420 to 1.3440
Quantity 500 mL
Beilstein 02, 18
Residue after Evaporation 0.0005% max.
Fieser 05,446
Merck Index 15, 6148
Specific Gravity 0.968
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
Viscosity 0.35 mPa.s (20°C)
Formula Weight 60.05
Percent Purity 98%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material Methyl formate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468300 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number414345000,414340025,414340050
Lot NumberA0433410 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number414345000,414340025,414340050
Lot NumberA0462832 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number414345000,414340025,414340050
Lot NumberA0461125 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number414345000,414340025,414340050
Lot NumberA0425751 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number414345000,414340025,414340050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl formate, 98%, for spectroscopy, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.