missing translation for 'onlineSavingsMsg'
Learn More
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals Product Code.: 10562841

Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals

Product Code. 10562841
1 L, Glass Bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10562841

Brand: Thermo Scientific Acros 295520010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexideShow MoreShow Less
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Acidity 0.04mg KOH/g max.
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 1 L
Flash Point 87°C
Packaging Glass Bottle
Physical Form Liquid at 20°C
Refractive Index 1.4770 to 1.4800
Assay Percent Range 99.8+%
Residue after Evaporation 0.003% max.
Specific Gravity 1.1
Linear Formula (CH3)2SO
Viscosity 1.987 mPa.s (25°C)
Beilstein 01, IV, 1277
Water 0.1% max.
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity ≥99.8%
Grade Extra Pure
Chemical Name or Material Methyl sulfoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468985 Certificate TypeCertificate of Analysis Date08/01/2025 Catalog Number295520010,295520250,295520025,295522500
Lot NumberA0450499 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number295520010,295520250,295520025,295522500
Lot NumberA0455409 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number295520010,295520250,295520025,295522500
Lot NumberA0453608 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number295520010,295520250,295520025,295522500
Lot NumberA0406934 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number295520010,295520250,295520025,295522500
Product Identifier
  • Methyl sulfoxide
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    17

    Minutes

    52

    Seconds

    55
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    60

    Hours

    6

    Minutes

    7

    Seconds

    4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.