missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade
£33.97 - £121.61
Chemical Identifiers
CAS | 60-11-7 |
---|---|
Molecular Formula | C14H15N3 |
Molecular Weight (g/mol) | 225.30 |
MDL Number | MFCD00008308 |
InChI Key | JCYPECIVGRXBMO-UHFFFAOYSA-N |
Synonym | 4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2 |
PubChem CID | 6053 |
ChEBI | CHEBI:17903 |
IUPAC Name | N,N-dimethyl-4-phenyldiazenylaniline |
SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10219183
|
Thermo Scientific Chemicals
151400250 |
25 g | Glass bottle |
£33.97
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10193720
|
Thermo Scientific Chemicals
151401000 |
100 g | Glass bottle |
£42.51
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10170030
|
Thermo Scientific Chemicals
151405000 |
500 g | Glass bottle |
£121.61
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
60-11-7 | |
225.30 | |
JCYPECIVGRXBMO-UHFFFAOYSA-N | |
6053 | |
N,N-dimethyl-4-phenyldiazenylaniline |
C14H15N3 | |
MFCD00008308 | |
4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2 | |
CHEBI:17903 | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Specifications
111.0°C to 117.0°C | |
C14H15N3 | |
MFCD00008308 | |
Solubility in water: 13.6 mg/l. Other solubilities: soluble ca. 140g/L in acetone, soluble in benzene, chloroform, diethyl ether,, ethanol, petroleum ether, vegetable oils, alcohol,, ether, strong mineral acids and soils | |
Authentic | |
N,N-dimethyl-4-phenyldiazenylaniline | |
6053 | |
225.29 | |
Pure | |
Methyl Yellow |
60-11-7 | |
(CH3)2NC6H4N=NC6H5 | |
4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2 | |
JCYPECIVGRXBMO-UHFFFAOYSA-N | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
225.30 | |
CHEBI:17903 | |
3% max. | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methyl Yellow, indicator grade
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Acute toxicity Category 3
Hazard Statement
- H351-Suspected of causing cancer.
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash hands and any exposed skin thoroughly after handling.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only