missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue hydrate, pure

£53.37 - £697.47

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9 hydrate, C.I. 52015 hydrate
PubChem CID 16211647
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10045700
View Documents Promotion Details
Thermo Scientific Chemicals
414251000
100 g Glass bottle
£53.37
100g
In Stock
to see stock.
Add to basket
 
10740102
View Documents
Thermo Scientific Chemicals
414255000
500 g Glass bottle
£188.94
500g
In Stock
to see stock.
Add to basket
 
11936931
View Documents
Thermo Scientific Chemicals
414250025
2.5 kg Plastic Drum
£697.47
2.50kg
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

122965-43-9
319.85
CXKWCBBOMKCUKX-UHFFFAOYSA-M
16211647
C16H18ClN3S
MFCD00150006
Basic Blue 9 hydrate, C.I. 52015 hydrate
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C

Specifications

100.0°C to 110.0°C
C16H18ClN3S
Basic Blue 9 hydrate, C.I. 52015 hydrate
CXKWCBBOMKCUKX-UHFFFAOYSA-M
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
319.85
319.85
≥70% (TiCl3-titration)
Green
Methylene Blue hydrate
122965-43-9
MFCD00150006
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol
Authentic
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
16211647
22% max.
Pure
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Warning

Product Identifier
  • Methylene Blue hydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Phenothiazin-5-ium,3,7-bis(dimethylamino)-, chloride, hydrate, Methylene Blue

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only