missing translation for 'onlineSavingsMsg'
Learn More
Methylmercury(II) hydroxide, 1M in H2O, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™ Product Code.: 15411707

Methylmercury(II) hydroxide, 1M in H2O, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15411707
10 mL
Click to view available options
Quantity:
1 mL
10 mL
50 mL
This item is not returnable. View return policy

Product Code. 15411707

Brand: Thermo Scientific Alfa Aesar 041253.AB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-57-2
MDL Number MFCD00013924

Specifications

Color Colorless
Physical Form Liquid
Quantity 10 mL
Linear Formula CH3HgOH
UN Number UN2024
Solubility Information Slightly soluble in water.
Formula Weight 232.63
Concentration or Composition (by Analyte or Components) 1M in H2O
Packaging ChemSeal™ bottles
Chemical Name or Material Methylmercury(II) hydroxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW26I012 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number041245.03,041245.14,041253.AB,041245.06
Lot NumberX05I020 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number041245.03,041245.14,041253.AB,041245.06
Lot NumberB05U033 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number041245.03,041245.14,041253.AB,041245.06
Lot NumberH07T034 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041245.03,041245.14,041253.AB,041245.06
Lot NumberA14N33 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041245.03,041245.14,041253.AB,041245.06
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methylmercury(II) hydroxide, 1M in H2O, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.