missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metolazone, Fisher BioReagents
£19.55 - £208.00
Chemical Identifiers
CAS | 17560-51-9 |
---|---|
Molecular Formula | C16H16ClN3O3S |
Molecular Weight (g/mol) | 365.83 |
MDL Number | MFCD00069304 |
InChI Key | AQCHWTWZEMGIFD-UHFFFAOYNA-N |
Synonym | metolazone, zaroxolyn, diulo, mykrox, microx, metalozone, metenix, oldren, metolazona, metolazonum |
PubChem CID | 4170 |
ChEBI | CHEBI:64354 |
SMILES | CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10728705
|
Fisher BioReagents
BP2667-25 |
25 mg | Amber Glass | N/A | ||||||
10596574
|
Fisher BioReagents
BP2667-50 |
50 mg | Amber Glass | N/A | ||||||
10134624
|
Fisher BioReagents
BP2667-100 |
100 mg | Amber Glass |
£208.00
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Metolazone is an anti-hypertensive.Chemical Identifiers
17560-51-9 | |
365.83 | |
AQCHWTWZEMGIFD-UHFFFAOYNA-N | |
4170 | |
CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O |
C16H16ClN3O3S | |
MFCD00069304 | |
metolazone, zaroxolyn, diulo, mykrox, microx, metalozone, metenix, oldren, metolazona, metolazonumShow More | |
CHEBI:64354 |
Specifications
17560-51-9 | |
White | |
MFCD00069304 | |
metolazone, zaroxolyn, diulo, mykrox, microx, metalozone, metenix, oldren, metolazona, metolazonum | |
CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O | |
365.83 | |
CHEBI:64354 | |
Powder or Solid |
248°C | |
C16H16ClN3O3S | |
15, 6224 | |
AQCHWTWZEMGIFD-UHFFFAOYNA-N | |
7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2-dihydroquinazoline-6-sulfonamide | |
4170 | |
365.8 | |
Metolazone |
Safety and Handling

Recommended Storage : RT
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Metolazone, Fisher BioReagents