Learn More
Emma Safety Footwear Metric Safety Shoes
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
Brand:  Emma Safety Footwear MM50654042
 View more versions of this product
Product Code. 15998541
  • £156.00 / 1 pair
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
White
Hydro-Tec™ Sanitized Silver
42
Elastic
Metric
Microfiber
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Emma Safety Footwear Metric Safety Shoes > Size: 42