Learn More
Metro™ Super Erecta™ Wire Shelving Accessory, Stem Casters
Choose from various types of casters to match your corrosion resistance needs
£43.25 - £307.00
Specifications
Diameter (English) | 5 in. |
---|---|
Diameter (Metric) | 12.7 cm |
For Use With (Equipment) | Metro™ Super Erecta™ wire shelving |
Alert:
The total weight of equipment and load should not exceed three times the load rating per caster.
Includes: Donut bumpers.
Product Code | Brand | Load Bearing Capacity (Metric) | Product Type | Material | Wheel Tread | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity (Metric) | Product Type | Material | Wheel Tread | Price | Quantity & Availability | |||
15986711
|
Metro™
5MDBGSA |
68 kg | Brake Stem Caster | Stainless Steel | High Modulus Donut |
£213.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15906721
|
Metro™
5MDGSA |
68 kg | Swiveled Stem Caster | Stainless Steel | High Modulus Donut |
£201.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15976731
|
Metro™
5PCB/Q-0299692-REV1 |
135 kg | Brake Stem Caster | Polymer | Polyurethane, Flat |
£112.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15966721
|
Metro™
5MHTPB |
135 kg | High-temperature Autoclave Brake Stem Caster | Stainless Steel | High-temp Phenolic |
£307.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15936721
|
Metro™
5MHTN |
135 kg | High-temperature Autoclave Swivel Stem Caster | Stainless Steel | High-temp Nylon |
£289.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15946721
|
Metro™
5MHTNB |
135 kg | High-temperature Autoclave Brake Stem Caster | Stainless Steel | High-temp Nylon |
£307.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15986731
|
Metro™
5PCBM |
135 kg | Brake Stem Caster | Antimicrobial Polymer | Polyurethane, Flat |
£121.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15996731
|
Metro™
5PCM |
135 kg | Swivel Stem Caster | Antimicrobial Polymer | Polyurethane, Flat |
£113.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15956721
|
Metro™
5MHTP |
135 kg | High-temperature Autoclave Swivel Stem Caster | Stainless Steel | High-temp Phenolic |
£289.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15966731
|
Metro™
5PC/Q-0299692-REV1 |
135 kg | Swivel Stem Caster | Polymer | Polyurethane, Flat |
£101.00
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
11705729
|
Metro™
5M |
90 kg | Swivel Caster | Steel | Resilient, Flat |
£43.25
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||
15956711
|
Metro™
5MB |
90 kg | Brake Stem Caster | Steel | Resilient, Flat |
£53.05
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15956731
|
Metro™
5MR |
90 kg | Rigid Stem Caster | Steel | Resilient, Flat |
£51.45
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15966711
|
Metro™
5MDA |
111 kg | Swivel Caster | Steel | High Modulus Donut |
£49.35
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||
15916731
|
Metro™
5MPR |
135 kg | Rigid Stem Caster | Steel | Polyurethane, Flat |
£74.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15976711
|
Metro™
5MDBA |
111 kg | Brake Stem Caster | Steel | High Modulus Donut |
£57.10
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15906731
|
Metro™
5MPGSA |
135 kg | Swivel Stem Caster | Stainless Steel | Polyurethane, Flat |
£206.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
15976721
|
Metro™
5MPBGSA |
135 kg | Brake Stem Caster | Stainless Steel | Polyurethane, Flat |
£216.00
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
Description
Caster choices include standard stem, polymer, stainless steel cart-washable and high-temperature autoclavable. Both polymer and stainless casters provide superior corrosion resistance, performance and enhanced durability. Casters available in swivel, swivel brake or rigid styles.
- Rigid stem casters are ordered two per unit for improved tracking and handling
- Rigid caster channels are provided with each two rigid stem casters and is based on shelf width
Polymer Casters
- Polymer casters offer corrosion resistance and enhanced durability
- All-polymer horn, stainless steel axle and hardware
- Polyurethane wheel
- Sleeve-style axle bearing
- Optional toe-operated brake mechanism
Stainless Steel Cart-Washable Casters
- Grease sealed with zerk fittings in swivel and axle
High-temperature Autoclave Casters
- Temperature rated for up to 300°F (149°C)
Order Info
Shelf width must be known when ordering rigid casters. Brakes are foot-operated.
Specifications
5 in. | |
12.7 cm | |
Metro™ Super Erecta™ wire shelving |