missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrode Fill Solutions

Made from the highest quality reagents to provide optimum performance.

Brand:  Mettler Toledo™ 51343184

9.54 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.


 View more versions of this product

Product Code. 10587335

  • £11.70 / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Aqueous solution containing inorganic and organic components
  • Supplied in 25mL (0.9 oz.) bottle

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.9 oz.
3 mol/L
KCl saturated with AgCl; 25mL
25 mL
Reference Electrode
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ Electrode Fill Solutions > KCl saturated with AgCl; 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.