missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff
GREENER_CHOICE

Disposable nitrile gloves with extended cuff provide durable protection in the event of chemical splashes. NFPA and medical exam grade certified for use by healthcare professionals.

£36.40

Specifications

Certifications/Compliance NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 455 1-4
Applications Healthcare, Laboratory
Color Black
Disposable Single-use
View More Specs

Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16499405
GREENER_CHOICE
View Documents Promotion Details
Ansell
93862070
Small
£36.40
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
16409415
GREENER_CHOICE
View Documents Promotion Details
Ansell
93862080
Medium
£36.40
Pack of 100
In Stock
to see stock.
Add to basket
 
16419415
GREENER_CHOICE
View Documents Promotion Details
Ansell
93862090
Large
£36.40
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
16429415
GREENER_CHOICE
View Documents Promotion Details
Ansell
93862100
X-Large
£36.40
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
16439415
GREENER_CHOICE
View Documents
Ansell
93862110
2X-Large
£36.40
Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
 
Description

Description

  • Tested against both fentanyl and gastric acid (vomit) to simulate authentic overdose situations
  • Dark, black color makes it easy to see when hands have come into contact with potentially hazardous powders
  • Made with a soft, flexible nitrile formulation
  • Fully textured for a reliable, consistent grip
  • For use with:
    • Administering drugs
    • Emergency services
    • Equipment repair and maintenance
    • Extra protection over wrist and forearm
    • Inspection, selecting, checking parts
    • Laboratory analysis and testing
    • Paint and spray shops
    • Patient examinations
    • Raw material sample collection
    • Tire rotation and change
Specifications

Specifications

NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600
Healthcare, Laboratory
Single-use
6.3 mil
Yes
4.7 mil
Powder Free
0.16 mm
12 in.
EN 420:2003 + A1:2009, EN 455 1-4
Black
Fully Textured
0.16 mm
Nitrile
0.12 mm
6.3 mil
MICROFLEX™ MidKnight™ XTRA 93-862
30 cm
Videos
SDS
Documents

Documents

Product Certifications