Learn More
Honeywell Safety Products™ Millenia Safety Glasses Product Code.: 11807000
In Stock
to see stock.
Add to basket
Honeywell Safety Products™ Millenia Safety Glasses
Product Code. 11807000
Black, Silver Mirror, Adjustable, breakaway Flexicord retainer, Impact-resistant, Scratch-resistant, UV-treated
Click to view available options
Frame Color:
Black
Blue
Silver
Lens Tint:
Clear
Silver Mirror
Yellow
Includes:
Adjustable, breakaway Flexicord retainer
Flexicord retainer
Lens Coating:
Impact-resistant, Anti-fog, UV-treated
Impact-resistant, Scratch-resistant, UV-treated
Product Code. 11807000
Brand: Honeywell Safety Products™ 1000004
In Stock
to see stock.
Add to basket
Add to basket
Virtually weightless eye protection for the active work environment. Honeywell Safety Products™ Millenia Safety Glasses block UV and provide protection for mechanical work including grinding, turning and milling.
  • General use and protection against impact (45m/s).
  • Suitable for mechanic work: grinding, turning, milling.
  • Polycarbonate lenses offer 99.9% UV protection up to 380nm.
  • Sensitive Tension Design and Parallel Vision Control.
  • A balanced fit and a greater visual comfort.
  • A virtually weightless feel.
  • Adjustable, breakaway Flexicord retainer.
  • Added security and convenience in the active work environment.
  • Unique 9 base curved lens design.
  • Excellent optical qualities and complete protection.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1131, EC Attestation Number EC1131
Lens Coating Impact-resistant, Scratch-resistant, UV-treated
Lens Tint Silver Mirror
Product Type Safety Glasses
For Use With (Application) Administration, Agriculture, Automotive, Aviation, Building and Construction, Energy or Electricity, Ship Building, Transportation, Utility and Wood industries.
Frame Color Black
Includes Adjustable, breakaway Flexicord retainer
No. per Pack 1 Ea.
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400
Product Title
Honeywell Safety Products™ Millenia Safety Glasses >