Learn More
Honeywell Safety Products™ Millenia Safety Glasses
Click to view available options
Frame Color:
Black
Blue
Silver
Lens Tint:
Clear
Silver Mirror
Yellow
Includes:
Adjustable, breakaway Flexicord retainer
Flexicord retainer
Lens Coating:
Impact-resistant, Anti-fog, UV-treated
Impact-resistant, Scratch-resistant, UV-treated
Description
Plano lens fashion safety spectacle, wrap around design with abrasion resistant lens and integral side shields. Available in a range of frame and lens colors.
Specifications
Specifications
Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1131, EC Attestation Number EC1131 |
Lens Coating | Impact-resistant, Scratch-resistant, UV-treated |
Lens Tint | Silver Mirror |
Product Type | Safety Glasses |
For Use With (Application) | Administration, Agriculture, Automotive, Aviation, Building and Construction, Energy or Electricity, Fishing, Logistics, Maintenance, Ship Building, Transportation, Utility and Wood industries |
Frame Color | Black |
Includes | Flexicord retainer |
No. per Pack | 1 Ea. |
Weight (Metric) | 32 g |
Lens Material | Polycarbonate |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Product Content Correction
Product Title
Honeywell Safety Products™ Millenia Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction