Learn More
ITW Chemische Produkte™ Mini AlphaMop™ AlphaWipe™ Replacement Mop Covers
Designed for use with the AlphaMop™ Isolator Cleaning Tool. Mini AlphaMop™ AlphaWipe™ Replacement Mop Covers are double bagged and are made of AlphaWipe™ polyester for effective cleaning with low contamination.
Brand: ITW Chemische Produkte™ TX7114E
This item is not returnable.
View return policy
Description
- Made of AlphaWipe™ polyester for effective cleaning and low contamination
- Cleanroom laundered for use in sterile and aseptic applications
- Designed to slip on and off easily, yet remain secure while in use
- Replacement mop pads available separately
Specifications
Mop Cover | |
AlphaMop Isolator Cleaning Tool | |
Double Bagged | |
Polyester |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
ITW Chemische Produkte™ Mini AlphaMop™ AlphaWipe™ Replacement Mop Covers > Replacement mop covers
Spot an opportunity for improvement?Share a Content Correction