Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™
Versatile check valves can be installed in any position
Brand: Cole-Parmer 50115-22
This item is not returnable.
View return policy
Description
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressureSpecifications
0.3125 in. | |
5/16 in. hose barb | |
Viton |
0.13 | |
Barbed |
Product Content Correction
Product Title
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™ > 5/16 in. hose barb
Spot an opportunity for improvement?Share a Content Correction