Learn More
Mitomycin C, Fisher BioReagents™
£108.00 - £241.00
Chemical Identifiers
CAS | 50-07-7 |
---|---|
Molecular Formula | C15H18N4O5 |
Molecular Weight (g/mol) | 334.332 |
InChI Key | NWIBSHFKIJFRCO-WUDYKRTCSA-N |
Synonym | mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c |
PubChem CID | 5746 |
ChEBI | CHEBI:27504 |
SMILES | CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10182953
|
Fisher BioReagents
BP2531-2 |
2 mg | Amber Glass |
£108.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10022604
|
Fisher BioReagents
BP2531-10 |
10 mg | Amber Glass |
£141.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115004
|
Fisher BioReagents
BP2531-20 |
20 mg | Amber Glass |
£241.00
20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mitomycin C
Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.Chemical Identifiers
50-07-7 | |
334.332 | |
mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c | |
CHEBI:27504 |
C15H18N4O5 | |
NWIBSHFKIJFRCO-WUDYKRTCSA-N | |
5746 | |
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N |
Specifications
50-07-7 | |
Blue | |
C15H18N4O5 | |
mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c | |
NWIBSHFKIJFRCO-WUDYKRTCSA-N | |
334.332 | |
CHEBI:27504 | |
99.0% min. (RP CH3CN:H20 0 to 100%), 98.0% min. (RP MEOH:H20 90:10) | |
Mitomycin C |
360°C | |
≥98.0 % | |
15, 6300 | |
Soluble in water | |
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N | |
5746 | |
334.3 | |
Solid |
Safety and Handling
WARNING!
Emergency Overview
Toxic if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is not required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:0
Instability:0
Recommended Storage : 4°C
- Mitomycin C
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /