missing translation for 'onlineSavingsMsg'
Learn More
Mitomycin C, Fisher BioReagents™ Product Code.: 10022604

Mitomycin C, Fisher BioReagents™

Product Code. 10022604
10 mg, Amber Glass
Click to view available options
Quantity:
2 mg
10 mg
20 mg
Packaging:
Amber Glass
This item is not returnable. View return policy

Product Code. 10022604

Brand: Fisher BioReagents BP253110

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mitomycin C

Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.

Recommended Storage: 4°C. Keep containers tightly closed in a dry, cool and well-ventilated place. Keep refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
Synonym mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Melting Point 360°C
Color Blue
Assay Percent Range ≥98.0 %
Packaging Amber Glass
Quantity 10 mg
Merck Index 15, 6300
Solubility Information Soluble in water
Formula Weight 334.3
Percent Purity 99.0% min. (RP CH3CN:H20 0 to 100%), 98.0% min. (RP MEOH:H20 90:10)
Physical Form Solid
Chemical Name or Material Mitomycin C
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Mitomycin C
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mitomycin C, Fisher BioReagents™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.