missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MitoProbe™ TMRM Assay Kit for Flow Cytometry

The MitoProbe TMRM Assay Kit contains tetramethylrhodamine methyl ester (TMRM) for the detection of mitochondrial membrane potential state and CCCP for the induction of mitochondrial membrane depolarization, if desired.

Brand:  Thermo Scientific™ M20036

144.72 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 15934294

  • £216.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

• Study mitochondrial membrane potential in live cells
• Quick, easy protocol

Determining the state of mitochondrial health can be achieved by detecting changes in the mitochondrial membrane potential. In a healthy cell with active mitochondria, TMRM is readily sequestered, thereby emitting a red-orange fluorescent signal. When apoptosis is induced or a treatment like CCCP is applied, the mitochondrial membrane is depolarized and the TMRM signal diminishes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

  • 1 vial TMRM (100 tests)
  • 1 vial 50 mM CCCP
  • Store at -5°C to -30°C.
TMRM Assay
Flow Cytometry Assay Kit
Vial(s)
Wet Ice
Fluorescence
Flow Cytometer
561
100 assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2983216 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberM20036
Lot Number2776001 Certificate TypeCertificate of Analysis Date20/12/2023 Catalog NumberM20036
Lot Number2738317 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog NumberM20036
Lot Number2633316 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog NumberM20036
Lot Number2539065 Certificate TypeCertificate of Analysis Date26/09/2022 Catalog NumberM20036
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MitoProbe™ TMRM Assay Kit for Flow Cytometry > Store at -5 to -30°C.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.