missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Poly-HRP

Amplify signal and improve detection of biotinylated antibodies for IHC and other methods using this streptavidin conjugated with HRP polymers.

Brand:  Thermo Scientific™ 21140

247.00 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.



Product Code. 10571164

  • £266.00 / 0.50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for IHC and other methods.

This poly-HRP conjugate is designed to deliver the highest sensitivity and low background in immunoassays where sample volume is limited or when the target molecule is present at low levels. Pierce Streptavidin Poly-HRP is purified to remove unconjugated streptavidin molecules that reduce signal intensity by competing for binding sites with horseradish peroxidase (HRP)-conjugates molecules. In addition, the conjugate is devoid of HRP monomers that can cause background signal. The Pierce Streptavidin Poly-HRP is compatible with chromogenic, fluorogenic and chemiluminescent HRP substrates used in ELISA, Western blotting, immunohistochemistry (IHC) and nucleic acid hybridization assays.

Highlights:

  • High sensitivity – detect low-abundant targets (low picogram-femtogram range) with high signal-to-noise ratios

  • Robust – consistent manufacturing and purification to eliminate low- and un-conjugated molecules for minimal background and high sensitivity

  • Flexible – compatible with chromogenic, fluorogenic and chemiluminescent substrates

  • Versatile – compatible with ELISA, Western blotting and immunohistochemistry (IHC)

  • Easy to use – can be directly substituted into immunoassays and other detection assays

  • Convenient – ready-to-use stabilized liquid format stored at 4°C

  • Cost effective – requires less conjugate per assay and standard HRP conjugates

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 mL
Stored at 4°C
HRP (Horseradish Peroxidase)
Western Blotting, ELISA, Immunohistochemistry
HRP (Horseradish Peroxidase)
Biotin
Biotin
Liquid
Pierce™
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL407179 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number21140
Lot NumberZL407179 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number21140
Lot NumberZJ401955 Certificate TypeCertificate of Analysis Date19/10/2024 Catalog Number21140
Lot NumberSPECS Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number21140
Lot NumberZI401955 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number21140
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    36

    Hours

    6

    Minutes

    58

    Seconds

    16
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Streptavidin Poly-HRP > Poly-HRP Conjugated; 0.5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.