missing translation for 'onlineSavingsMsg'
Learn More
ML 190, Tocris Bioscience™ Product Code.: 30002023

ML 190, Tocris Bioscience™

Product Code. 30002023
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30002023

Brand: Tocris Bioscience 4866/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective κ opioid receptor antagonist

Selective κ opioid receptor (KOP) antagonist (IC50 = 120 nM in a β-arrestin assay); displays >267-fold selectivity over μ and δ opioid receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1355244-02-8
Molecular Formula C27H32N6O3
Molecular Weight (g/mol) 488.592
InChI Key PMTIWRPLQBVEMR-UHFFFAOYSA-N
Synonym n-3-4-4-methoxyphenyl piperazin-1-yl propyl-2-1-methyl-6-oxopyrido 2,3-e pyrrolo 1,2-a pyrazin-5 6h-yl acetamide, n-3-4-4-methoxyphenyl-1-piperazinyl propyl-1-methyl-6-oxopyrido 2,3-e pyrrolo 1,2-a pyrazine-5 6h-acetamide, n-3-4-4-methoxyphenyl piperazin-1-yl propyl-2-13-methyl-7-oxo-2,8,10-triazatricyclo 7.4.0.0 2 ,? trideca-1 13 ,3,5,9,11-pentaen-8-yl acetamideShow MoreShow Less
PubChem CID 44665680
SMILES CC1=C2C(=NC=C1)N(C(=O)C3=CC=CN32)CC(=O)NCCCN4CCN(CC4)C5=CC=C(C=C5)OC

Specifications

Quantity 50 mg
Formula Weight 488.58
Chemical Name or Material ML 190
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ML 190, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.