Learn More
SymLine Module Signal Set (EU)
Monitor two filling levels at once, perfect integration into lab furniture with the Switch Amplifier
Brand: SymLine 106678
545.13 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Connect NAMUR-Sensors or potential-free mechanical contacts which are mounted in the zones 0, 1, 2 (Gas) or 20, 21, 22 (dust)
- Compact design
Specifications
Module Signalset for safety cabinets 'EU-plug', consisting of: 1 x Switch amplifier EU, 2 channels (108278) 1 x Housing for the isolating switch amplifier (106590) 2 x Cable set: Connection for switch amplifier - Built-in signal box (108314) | |
Safety Cabinets | |
Module Signal Set (EU) | |
1 x Switch amplifier EU, 2 channels (108278) 1 x Housing for the isolating switch amplifier (106590) 2 x Cable set: Connection for switch amplifier - Built-in signal box (108314) |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
SymLine Module Signal Set (EU) > Type: Switch Amplifier; Module Signalset
Spot an opportunity for improvement?Share a Content Correction