Learn More
Invitrogen™ Albumin from Bovine Serum (BSA), Alexa Fluor™ 647 conjugate
Suited for applications in which well-defined molecular weight tracers are needed
Brand: Invitrogen™ A34785
234.50 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
- Applications include studying the rate of endocytosis and exocytosis and following intracellular protein processing
Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature

Specifications
5 mg | |
Bovine | |
Alexa Fluor Dyes | |
Lyophilized | |
Alexa Fluor™ |
Alexa Fluor 647 | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Albumin Proteins |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
31Hours
22Minutes
3Seconds
48New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.