missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calcium Green™-1 dextran, Potassium Salt, 3000 MW, Anionic

Product Code. 11588946
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11588946

Brand: Invitrogen™ C6765

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+

  • Uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues
  • Cells may be physically loaded with the cell-impermeant salt forms of these dextran-conjugated indicators using patch pipet, microinjection, or Influx™ pinocyte-loading reagent
  • Fluorescence signal from these cells is measured using fluorescence microscopy
  • Dextran forms of calcium indicators show a dramatic reduction in both leakage and compartmentalization compared to the AM ester forms
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • These probes are excited by visible light, and because the energy required for excitation is low, the potential for cellular photodamage is reduced
  • Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem

  • Label (Ex/Em): Calcium Green-1 (506/531nm)
  • Fluorescence intensity increase upon binding Ca2+: approximately 14 fold

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Product Line Calcium Green
Quantity 5 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.