missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Low Density Lipoprotein from Human Plasma, DiI complex (DiI LDL)

Nonacetylated conjugate

Brand:  Invitrogen™ L3482

421.04 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10760194

  • £560.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Human low-density lipoprotein (LDL) is large protein complex (approx. 500,000 Da) that binds to a specific receptor on surface of vertebrate cells and delivers cholesterol via receptor-mediated endocytosis. Experiments to study LDL are typically performed by adding fluorescently labeled LDL to cultured cells and analyzing them by microscopy or flow cytometry. Alternatively fluorescently labeled LDL can be injected into test animals, and distribution of the label can be analyzed after the specified time period.

  • Label (Ex/Em): Dil (554/571)
  • Acetylated: No
  • Amount: 200μL (1.0mg/mL
  • Nonacetylated LDL products are shipped within two weeks of their preparation
  • LDL containing unmodified apoprotein is used to study normal cholesterol delivery and internalization
  • Applications for labeled LDL complexes include: Counting cell-surface LDL receptors, and analyzing their motion and clustering following internalization; Quantitating LDL receptor activity in fibroblasts (replacing the radiolabeled LDL assay); Investigating LDL expression and identifying LDL receptor deficiencies in various cell lines

Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mg⁄ml
Fluorescence
Other Label(s) or Dye(s)
200 μL
Store in refrigerator (2–8°C) and protect from light.
Supplement
Liquid
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3011684 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberL3482
Lot Number2831377 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog NumberL3482
Lot Number2626538 Certificate TypeCertificate of Analysis Date04/08/2023 Catalog NumberL3482
Lot Number2540902 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog NumberL3482
Lot Number2478008 Certificate TypeCertificate of Analysis Date06/04/2022 Catalog NumberL3482
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Low Density Lipoprotein from Human Plasma, DiI complex (DiI LDL) > 200μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.