Learn More
Molybdenum sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Description
Molybdenum sputtering target is used as resistance heating elements in furnaces, as filament supports in electric lamps, and as electrodes for mercury vapor lamps. It is also used as an alloying agent in certain grades of steel, Permalloys and Stellites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 Ea. |
CAS | 7439-98-7 |
Molecular Formula | Mo |
MDL Number | MFCD00003465 |
Synonym | molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 |
Solubility Information | Insoluble in water. Soluble in nitric acid and sulfuric acid. Slightly soluble in hydrochloric acid. |
InChI Key | ZOKXTWBITQBERF-UHFFFAOYSA-N |
SMILES | [Mo] |
IUPAC Name | molybdenum |
Molecular Weight (g/mol) | 95.95 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M15K031 | Certificate of Analysis | 05/03/2024 | 040866.KS |
W16I019 | Certificate of Analysis | 23/09/2022 | 040866.KS |
B07S059 | Certificate of Analysis | 28/12/2021 | 040866.KS |
C09S029 | Certificate of Analysis | 08/12/2021 | 040866.KS |
C12N33 | Certificate of Analysis | 08/12/2021 | 040866.KS |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.