missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Mono Q PC, Columns Prepacked with MonoBeads™
Use Mono Q or S precision columns for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale. Cytiva Life Sciences™ Mono Q or S PC, Columns Prepacked with MonoBeads™ achieve maximum resolution from the efficiency of small, perfectly spherical monodispersed particles.
Brand: Cytiva 17-5166-01
This item is not returnable.
View return policy
Description
- Maximum resolution achieved from efficiency of small spherical particles, optimally packed
- Particles are coupled to Q or S ion exchangers with excellent sensitivities
- Compatible with high-performance LC systems
- Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid
- Tricorn columns are optimized for the polishing step when high purity is required
- PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
- Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
- Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL
Specifications
All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid | |
1mL | |
5mm | |
1.96 in. | |
0.05 to 3mL/min. | |
O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3 | |
2 to 12 | |
Monodisperse porous Polystyrene/Divinyl Benzene bead | |
Mono Q Ion Exchange Column |
Mono Q ion exchange column | |
0.19 in. | |
Each | |
50mm | |
Borosilicate Glass | |
40 bar | |
10μm | |
4° to 30°C | |
Small-scale polishing in purification of Proteins, Peptides and other biomolecules when high purity is required |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Mono Q 5/50 GL
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol