missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Mono Q PC, Columns Prepacked with MonoBeads™
Use Mono Q or S precision columns for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale. Cytiva Life Sciences™ Mono Q or S PC, Columns Prepacked with MonoBeads™ achieve maximum resolution from the efficiency of small, perfectly spherical monodispersed particles.
£1510.00 - £7600.00
Specifications
Buffer | All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid |
---|---|
Quantity | Each |
Ligand Type | O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3 |
pH Range | 2 to 12 |
Particle Size | 10μm |
Product Code | Brand | Height (Metric) | Material | Max. Pressure | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Material | Max. Pressure | Price | Quantity & Availability | |||
10126094
|
Cytiva
17-0671-01 |
50mm | Glass | 50 bar | N/A | ||||
10146194
|
Cytiva
17-5179-01 |
100mm | PEEK | 40 bar | N/A | ||||
10402035
|
Cytiva
17-5167-01 |
100mm | Borosilicate Glass | 40 bar | N/A | ||||
10511280
|
Cytiva
17-5166-01 |
50mm | Borosilicate Glass | 40 bar | N/A | ||||
Description
- Maximum resolution achieved from efficiency of small spherical particles, optimally packed
- Particles are coupled to Q or S ion exchangers with excellent sensitivities
- Compatible with high-performance LC systems
- Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid
- Tricorn columns are optimized for the polishing step when high purity is required
- PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
- Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
- Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL
Specifications
All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid | |
O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3 | |
10μm | |
4° to 30°C |
Each | |
2 to 12 | |
Monodisperse porous Polystyrene/Divinyl Benzene bead | |
Mono Q Ion Exchange Column |