Learn More
MP Biomedicals™ AIN-93G Mineral Mix, Powder
Powdered, for rodents
Brand:  MP Biomedicals™ 0296040010
 View more versions of this product
Product Code. 11481442
  • £382.00 / 10kg
Explore more special offers
Description
Description
For growth, pregnancy and lactational phases of rodents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Powder
10 kg
15 to 30°C
Calcium Carbonate 35.7%, Monopotassium Phosphate 19.6%, Potassium Citrate Monohydrate 7.078%, Sodium Chloride 7.4%, Potassium Sulfate 4.66%, Ma gnesium Oxide 2.4%, Ferric Citrate 0.606%, Zinc Carbonate 0.165%, Man ganese Carbonate 0.063%, Copper Carbonate 0.03%, Potassium Iodate 0.001%, Sodium Selenate, Anhydrous 0.00103%, Ammonium Molybdate.4H2O 0.000795%, Sodium Metasilicate.9H2O 0.145%, Chromium Potassium Sulfate.12H2O 0.0275%, Lithium Chloride 0.00174%, Boric Acid 0.008145%, Sodium Fluoride 0.00635%, Nickel Carbonate 0.00318%, Ammonium Vanadate 0.00066%, Powdered Su gar 22.1%
Growth, pregnancy and lactational phases of Rodents
Rodent
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.