Learn More
MP Biomedicals™ AIN-93M Mineral Mix, Powdered
AIN-93M Mineral Mix, Powdered, for rodents.
Brand:  MP Biomedicals™ 0296040102
 View more versions of this product
Product Code. 11401452
  • £85.15 / 2kg
Explore more special offers
Description
Description
  • Used for maintenance of adult rodents
  • Not available in pellet form
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Powder
2 kg
15°C to 30°C
Calcium Carbonate 35.7%, Monopotassium phosphate 19.6%, Potassium Citrate monohydrate 7.078%, Sodium Chloride 7.4%, Potassium Sulfate 4.66%, Ma gnesium Oxide 2.4%, Ferric Citrate 0.606%, Zinc Carbonate 0.165%, Man ganese Carbonate 0.063%, Copper Carbonate 0.03%, Potassium Iodate 0.001%, Sodium Selenate, Anhydrous 0.00103%, Ammonium Molybdate.4H2O 0.000795%, Sodium Metasilicate.9H2O 0.145%, Chromium Potassium Sulfate.12H2O 0.0275%, Lithium Chloride 0.00174%, Boric Acid 0.008145%, Sodium Fluoride 0.00635%, Nickel Carbonate 0.00318%, Ammonium Vanadate 0.00066%, Powdered Su gar 22.1%
Diet for maintenance of adult Rodent
Rodent
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ AIN-93M Mineral Mix, Powdered > AIN-93M Mineral Mix, Powdered; Quantity: 2 kg
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.