missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastPROTEIN™ Red Matrix

FastPROTEIN™ Red Matrix Kit designed for the rapid extraction of proteins from yeast cells, fungi and algea in a few seconds.

Brand:  MP Biomedicals™ 116550600

 View more versions of this product

Product Code. 11472400

  • £316.00 / Pack of 50
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket

Requires : FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin 2.0 mL tubes, Microcentrifuge tubes (2.0 mL and 1.5 mL)
This item is not returnable. View return policy

Description

Description

  • Kit or Assay Type: Protein Isolation, Mini-prep
  • Molecule to be Enriched: Proteins
  • Sample Type: Yeast and Yeast Spores
  • Tube Size: 2 mL
  • Matrix Composition: Matrix C, 1.0 mm silica spheres
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yeast breaking buffer
50 Preps
15°C to 30°C
Rapid isolation and purification of Protein from Yeast cultures
Red matrix kit
Matrix C, 1 mm Silica Spheres, Yeast breaking Buffer, Tubes
2 mL
FastProtein Red Matrix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.