missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ FastPROTEIN™ Red Matrix
FastPROTEIN™ Red Matrix Kit designed for the rapid extraction of proteins from yeast cells, fungi and algea in a few seconds.
£311.64 - £429.30
Specifications
Buffer | Yeast breaking buffer |
---|---|
Product Type | Red matrix kit |
Includes | Matrix C, 1 mm Silica Spheres, Yeast breaking Buffer, Tubes |
Content And Storage | 15°C to 30°C |
Capacity | 2 mL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482400
|
MP Biomedicals™
116550700 |
100 Preps |
£429.30
Pack of 100 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
11472400
|
MP Biomedicals™
116550600 |
50 Preps |
£311.64
Pack of 50 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
- Kit or Assay Type: Protein Isolation, Mini-prep
- Molecule to be Enriched: Proteins
- Sample Type: Yeast and Yeast Spores
- Tube Size: 2 mL
- Matrix Composition: Matrix C, 1.0 mm silica spheres
Specifications
Yeast breaking buffer | |
Matrix C, 1 mm Silica Spheres, Yeast breaking Buffer, Tubes | |
2 mL | |
FastProtein Red Matrix |
Red matrix kit | |
15°C to 30°C | |
Rapid isolation and purification of Protein from Yeast cultures |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.