Learn More
MP Biomedicals™ FastRNA™ Pro Red Kit, 2 mL Tubes, 50 Preps
FastRNA™ Pro Red Kit is used for total RNA isolation from up to 109 yeast and fungi cells. Uses Lysing Matrix C for cell lysis. For use with the FastPrep-24™ or FastPrep™ FP120 Instrument.
Brand: MP Biomedicals™ 116035050
263.32 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Requires : FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, RNase Erase™, Recommended: Chloroform, 100% ethanol, 75% ethanol, 1.5 or 2.0 mL RNase-free microcentrifuge tubes, Agarose, Gel loading dye and RNA size marker
This item is not returnable. View return policy
Description
- Product Overview: The FastRNA™ Pro Red Kit is designed to efficiently isolate total RNA from yeast and fungi. Using the FastPrep-24™ or FastPrep™ FP120 Instrument, up to 109 cells are homogenized by Lysing Matrix C in impact-resistant 2 mL tubes. Total RNA is released into the proprietary, protective RNApro™Solution. Following extraction with chloroform and precipitation, total RNA is extracted.
- Applications: FastRNA™ Pro Red Kit is used to isolate total RNA from yeast and fungi. Isolated RNA is suitable for RT-PCR analysis and other downstream applications.
- Sample Type: Spores, Fungi, Yeast, Algae
- Sample Volume: Up to 500 mg per prep

Specifications
Spores, Fungi, Yeast | |
RNApro™ Solution (1 x 55 mL Bottle), DEPC-H2O (1 x 15 mL Bottle), Lysing Matrix C (50 x 2 mL Tubes), Short Protocol, User Manual, MSDS, Certificate of Analysis | |
FastRNA-24 Kits |
RT-PCR analysis and other downstream applications | |
50 Preps | |
50 x 2 mL |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- RNApro Solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Phenol, Guanidine Thiocyanate, Glycerol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.