Learn More
Thermo Scientific™ Multidrop™ Combi Accessories

Guarantee excellent dispensing results and day-to-day performance with combi accessories.
£26.45 - £2768.39
Product Code | Brand | Description | Product Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Product Type | Price | Quantity & Availability | ||||||
10256343
|
Thermo Scientific™
24073290 |
Small tube plastic tip dispensing cassette | Tube Dispensing Cassette |
£624.37
Each |
In Stock
Log in to see stock. ![]() |
||||||
11590745
|
Thermo Scientific™
2805980 |
Tube Holder Assembly | N/A | N/A | N/A | ||||||
11580745
|
Thermo Scientific™
15280030 |
High version | Priming Vessel | N/A | N/A | N/A | |||||
14811252
![]() |
Thermo Scientific™
N13132 |
Small long-tube cassette, 5-pack | Tube Reagent Filter |
£338.00
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
11762038
|
Thermo Scientific™
24073291 |
Small tube plastic tip dispensing cassette 5-pack | Tube Dispensing Cassette |
£2768.39
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
11570735
|
Thermo Scientific™
24073293 |
Long small tube plastic tip dispensing cassette (tubing length 200cm) | Tube Dispensing Cassette |
£679.74
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
12101740
![]() |
Thermo Scientific™
24073295 |
Small tube metal tip dispensing cassette | Tube Dispensing Cassette |
£1020.00
Each |
In Stock
Log in to see stock. ![]() |
||||||
10095273
|
Thermo Scientific™
24073296 |
Small tube metal tip dispensing cassette 5-pack | Tube Dispensing Cassette |
£2755.08
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
11580735
|
Thermo Scientific™
24073298 |
Long small tube metal tip dispensing cassette (tubing length 200cm) | Tube Dispensing Cassette |
£679.74
Each |
In Stock
Log in to see stock. |
||||||
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi Accessories.
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi Accessories.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.