Learn More
Thermo Scientific™ Multidrop™ DW Reagent Dispenser
Accelerate larger scale assays with the superior precision and accuracy of this high-speed reagent dispenser designed for repetitive dispensing of large volumes.
Brand: Thermo Scientific™ 5840177
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Accelerate larger scale assays with the superior precision and accuracy of the Thermo Scientific™ Multidrop™ DW Reagent Dispenser, a high-speed reagent dispenser designed for repetitive dispensing of large volumes in pharmaceutical and biotechnology laboratories.
Features:
- Portions various liquids into 96 standard and DW plates across 2a 0 to 995μL volume range
- Precise dispensing ensures high-quality assay results and reproducible data
- Wide volume range provides flexibility and increased speed
- Quick plate and volume selection combined with easy maintenance make assay setup quick and easy
- Dispenser uses an eight-channel detachable and autoclavable dispensing cassette to ensure sterile conditions and to avoid cross contamination
- All reagent lines can be backflushed into the reagent bottle, minimizing the loss of expensive reagents
Includes:
One standard tube dispensing cassette (Cat. No. 24072670)
Recommended for:
- Cell based assays
- Compound storage
- Genomics and proteomics research
- ELISA assays
- Microbiological tests
Specifications
Reagent Dispenser | |
8 | |
5 μL | |
96-well plates; 96-deep well plates; 1.1mL tubes in 96 tube rack; Micronic 96-place tube racks | |
96-well plate: 20µL in 5 seconds; 50µL in 8 seconds; 300µL in 26 seconds; 900 µL in 74 seconds | |
12.2 in. | |
7.1 in. | |
12.6 in. | |
13.7 lb. | |
100 to 120V 50/60Hz | |
50/60 Hz |
20µLl: ±2% (typical); 100µL: ±1% (typical); 900µL: ±1% (typical) | |
20 to 995 μL | |
RS-232 | |
20μL: CV≤1.5% (typical), 100μL: CV≤1% (typical), 900μL: CV≤0.5% (typical) | |
<7 mL | |
31 cm | |
18 cm | |
32 cm | |
6.2 kg | |
100/120 V |