Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Modules
Automated reagent dispenser for 6- to 1536-well microplates.
Brand: Agilent BioTek 7210010
This item is not returnable.
View return policy
Description
- Automated reagent dispenser for 6- to 1536-well microplates
- Parallel Dispense technology allows up to four independent reagents to be dispensed in parallel without potential carryover
- RAD technology for dispensing to individual wells
- Cell friendly angled dispense and/or wash tubes won't disrupt cell layers
Applications:
- For benchtop and integrated automation workflows
- Bulk reagent dispensing
- Cell seeding
- Cell washing /media exchange
- Bead washing
Specifications
Peristaltic Pump Dispenser Module | |
MultiFlo FX Microplate Dispenser | |
Plastic tip cassette | |
1/Cs. |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Agilent BioTek MultiFlo FX Microplate Dispenser Modules > Adds 1 peristaltic pump dispenser
Spot an opportunity for improvement?Share a Content Correction