Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Modules
GREENER_CHOICE
Automated reagent dispenser for 6- to 1536-well microplates.
Brand:  Agilent BioTek 7210009
 View more versions of this product
Product Code. 12598816
  • £6550.00 / Each
Explore more special offers
Description
Description
  • Automated reagent dispenser for 6- to 1536-well microplates
  • Parallel Dispense technology allows up to four independent reagents to be dispensed in parallel without potential carryover
  • RAD technology for dispensing to individual wells
  • Cell friendly angled dispense and/or wash tubes won't disrupt cell layers
Applications:
  • For benchtop and integrated automation workflows
  • Bulk reagent dispensing
  • Cell seeding
  • Cell washing /media exchange
  • Bead washing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dual Syringe Drive Dispenser (Non-Auto)
MultiFlo FX Microplate Dispenser
1/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Agilent BioTek MultiFlo FX Microplate Dispenser Modules > Adds 2 syringe drive dispensers; Non-autoclavable